missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega PureYield™ Plasmid Maxiprep System
The PureYield Plasmid Maxiprep System isolates transfection-quality plasmid DNA. Yields up to 1mg of plasmid DNA from a 250ml bacterial culture.
Supplier: Promega A2392
Includes: All systems include reagents and columns for purification. A6730 also includes vacuum pump, vacuum manifold, and vacuum elution device.
Description
- Up to 1mg of Transfection-Ready Plasmid from 250mL Cultures
- One-hour, vacuum-based protocol
- Highly pure, concentrated plasmid DNA
- No tedious, gravity-drip columns or post-elution alcohol precipitation
Chemical Identifiers
| 15°C to 30°C | |
| Vac-Man™ laboratory vacuum manifold | |
| 250mL |
| Vacuum | |
| Isolates transfection-quality plasmid DNA | |
| Up to 1mg |
Specifications
| 15°C to 30°C | |
| Vac-Man™ laboratory vacuum manifold | |
| 130mL Neutralization solution, 2 × 5 PureYield clearing columns, 2 × 5 PureYield maxi binding columns, 36.5mL Endotoxin removal wash, 82mL Column wash, 125mL Cell resuspension solution, 125mL Cell lysis solution, 25mL Nuclease-free water | |
| Plasmid Maxiprep System | |
| Up to 1mg |
| Vacuum | |
| Isolates transfection-quality plasmid DNA | |
| 10 Preps | |
| 250mL |