missing translation for 'onlineSavingsMsg'
Learn More

Promega ProTEV Plus Protease

Catalog No. PRV6101
Encompass
Change view
Click to view available options
Quantity:
1000 U
8000 U
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV6101 1000 U
PRV6102 8000 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV6101 Supplier Promega Supplier No. V6101

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ProTEV Plus is an improved 48kDa version of the Nla protease from tobacco etch virus (TEV) that has been engineered to be more stable than native TEV protease for prolonged enzymatic activity.

  • Engineered Protease with Greater Stability, Prolonged Activity Over Native TEV Protease
  • Active over broad pH (5.5-8.5) and temperature (4-30°C) range
  • Cleaves protein fusions with the ENLYFQ(G/S) sequence
  • Use with in-solution and on-column digestions
  • Supplied at a concentration of 5u/μL

Specifications

Concentration 5 U/μL
For Use With (Application) Cleavage of affinity tags from fusion Proteins after Protein purification, removal of HaloTag™ Protein during Protein purification/expression of used Proteins of interest
Kit Contents HQ tag located at N-terminus to be immobilized on affinity resins
Content And Storage -30°C to -10°C
Quantity 1000 U
Product Type Protease