missing translation for 'onlineSavingsMsg'
Learn More

Promega Proteasome-Glo™ Cell-Based Assays

Catalog No. p-3309197
Encompass
Change view
Click to view available options
Quantity:
10 mL
5 x 10 mL
Product Type:
Chymotrypsin-like Cell-based Assay
Trypsin-like Cell-based Assay
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
PRG8660 10 mL Chymotrypsin-like Cell-based Assay
PRG8661 5 x 10 mL Chymotrypsin-like Cell-based Assay
PRG8760 10 mL Trypsin-like Cell-based Assay
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PRG8660 Supplier Promega Supplier No. G8660
Only null left

The Proteasome-Glo™ Cell-Based Assays are homogeneous, luminescent assays that measure the chymotrypsin-like, trypsin-like and caspase-like protease activities associated with the proteasome complex in cultured cells.

  • Homogeneous, Luminescent Assays that Measure Protease Activities in Cultured Cells
  • Simple add-mix-measure protocol
  • Chymotrypsin-Like, Trypsin-Like and Caspase-Like activity data directly from cellular environment
  • Results within 10-30 minutes after reagent addition

  • U.S. Pat. No. 6,602,677, Australian Pat. No. 754312 and other patents and patents pending.
  • The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713; and 5,700,673.

Specifications

Product Type Chymotrypsin-like Cell-based Assay
Includes 1 x 50μL Suc LLVY Glo substrate, 1 x 10mL Proteasome Glo Cell based Buffer, 1 x 1 each Luciferin detection Reagent
pH 7.2
Content And Storage -30°C to -10°C
Quantity 10 mL
Form Liquid