Learn More
Promega Proteasome-Glo™ Bioluminescent Enzyme Assays DFS Item
The Proteasome-Glo Assays are homogeneous, luminescent assays that individually measure the chymotrypsin-like, trypsin-like and caspase-like protease activities associated with the proteasome in a purified enzyme-based format (not suitable for cell-based applications).
Supplier:  Promega G8622
 View more versions of this product
Catalog No. PRG8622
Only null left
Explore available promotions
Description
Description
  • Measure Chymotrypsin-, Trypsin- and Caspase-Like Activities
  • Homogeneous, luminescent enzyme-based assays
  • Simple add-mix-measure protocol
  • Results in 10-30 minutes after reagent addition
Specifications
Specifications
Chymotrypsin-like Assay
Bioluminescence
-30°C to -10°C
50 mL
1 x 50mL Proteasome Glo Buffer, 1 x 250μL Suc LLVY Glo substrate, 1 x 1 each Luciferin detection Reagent
7.2
Proteasome-Glo
Liquid
Product Suggestions
Product Suggestions