missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega Proteasome-Glo™ Bioluminescent Enzyme Assays
Description
- Measure Chymotrypsin-, Trypsin- and Caspase-Like Activities
- Homogeneous, luminescent enzyme-based assays
- Simple add-mix-measure protocol
- Results in 10-30 minutes after reagent addition
Specifications
Specifications
| Product Type | Caspase-like Assay |
| Includes | 1 x 10mL Proteasome Glo Buffer, 1 x 50μL Z nLPnLD Glo substrate, 1 x 1 each Luciferin detection Reagent |
| Detection Method | Bioluminescence |
| pH | 7.2 |
| Content And Storage | -30°C to -10°C |
| Product Line | Proteasome-Glo |
| Quantity | 10 mL |
| Form | Liquid |