missing translation for 'onlineSavingsMsg'
Learn More

Promega PKCι Kinase Enzyme System

Catalog No. PRV9751
Encompass
Change view
Click to view available options
Quantity:
1/Ea.
10 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV9751 1/Ea.
PRV3751 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV9751 Supplier Promega Supplier No. V9751

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Recombinant full-length human PKC iota was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. Protein kinase C iota is a member of the protein kinase C family of serine-threonine kinases.

  • Easily Screen and Profile PKCι Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Specifications

Includes 1 x 10μg PKCl Kinase Enzyme system, 1 x 1000 ADP-Glo™ Kinase assay
Molecular Weight (g/mol) 98
Content And Storage Components at different temps
Quantity 1/Ea.
Form Fluid