missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega PKCδ Kinase Enzyme System
Description
- Easily Screen and Profile PKCδ Kinase Inhibitors
- Includes kinase, substrate and reaction buffer
- Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Specifications
| Includes | 1 x 10μg PKCd Kinase Enzyme system, 1 x 1000 ADP-Glo™ Kinase assay |
| Molecular Weight (g/mol) | 104 |
| Content And Storage | Components at different temps |
| Quantity | 1/Ea. |
| Form | Fluid |