missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega NanoLuc™ pNL3 Vectors
Description
- Vector that Expresses NanoLuc(R) Luciferase in the Presence of NF-κB
- Responds to changes in NF-κB expression in cells
- Includes an optimized NanoLuc(R) luciferase gene that codes for a reporter protein with a shorter half-life (NlucP)
- Hygromycin resistance selectable marker means choosing transient transfection or generating a stable cell line
Specifications
Specifications
| Product Type | pNL3.2.NF-κB-RE(NlucP/NF-κB-RE/Hygro) |
| Concentration | 1 μg/μL |
| pH | 8 |
| Content And Storage | -30°C to -10°C |
| Quantity | 20 μg |
| Form | Liquid |