missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega Modifying Enzymes: Single-Stranded DNA Binding Protein
Description
- Cooperatively Binds ssDNA with High Affinity
- Consists of four identical 18.9kDa subunits
- Does not bind well to double-stranded DNA
- Supplied at a concentration of 1-5μg/μL
Specifications
Specifications
| Concentration | 1 to 5 μg/μL |
| Color | Colorless |
| Components | 4 Identical 18.9kDa subunits |
| pH | 7.4 |
| For Use With (Application) | Studying DNA replication and recombination, DNA sequencing |
| Storage Buffer | 50mM Tris HCl (pH 7.4), 1mM EDTA, 1mM DTT, 0.2M NaCl, 50% Glycerol |
| Content And Storage | -30°C to -10°C |
| Source | E. coli |
| Quantity | 100 μg |
| Product Type | Single-stranded DNA Binding Protein |
| Show More |