missing translation for 'onlineSavingsMsg'
Learn More

Promega Maxwell™ 16 Purification Kits

Catalog No. p-3099136
Encompass
Change view
Click to view available options
Product Type:
Blood DNA Kit
LEV Blood DNA Kit
Tissue DNA Kit
Viral Total Nucleic Acid Kit
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
PRAS1010 Blood DNA Kit
PRAS1030 Tissue DNA Kit
PRAS1150 Viral Total Nucleic Acid Kit
PRAS1290 LEV Blood DNA Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. PRAS1010 Supplier Promega Supplier No. AS1010

Includes

All kits include 48 Maxwell kit-specific cartridges, 50 purification plungers, 50 elution tubes, and 20mL elution buffer. FFPE Tissue LEV DNA Purification Kit also includes 32mL lysis buffer, 2 x 10mg proteinase K, and 15mL incubation buffer. Viral Total Nucleic Acid Kit also includes 20mL lysis buffer, 2 x 1mL proteinase K solution, and 20mL nuclease-free water. LEV Blood DNA Kit also includes 20mL lysis buffer and 2 x 1mL proteinase K solution.

Requires

Maxwell 16 DNA Purification Instrument (Mfr. No. AS1120 not for use with Maxwell 16 MDx); LEV Kits also require the LEV Hardware Kit (Mfr. No. AS1250)

Used with Maxwell 16 Instruments to provide an easy method for efficient, automated purification of genomic DNA

  • Automated Extraction of DNA from Blood or Buffy Coat Samples
  • Processes up to 400μL of whole blood
  • For use with the Maxwell(R) 16 Instrument

  • U.S. Patents 6,027,945 and 6,368,800; Australian Pat. No. 732756.
  • Mfr. Nos. AS1010, AS1020, AS1030, and AS1140: This product is licensed for use under U.S. Patent 6,174,704.
  • Mfr. Nos. AS1130, AS1140: U.S. Patent 6,673,631.
  • Mfr. Nos. AS1130, AS1140: Patent pending.

Specifications

Content And Storage 15°C to 30°C
For Use With (Equipment) Maxwell 16 and 16 MDx instruments
For Use With (Application) Automated genomic DNA purification from whole blood and buffy coat samples
Includes 48 Maxwell™ 16 Blood DNA cartridges, 50 purification plungers, 50 Elution tubes, 20mL Elution Buffer
No. of Reactions 48 Preps
Product Type Blood DNA Kit