missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega Maxwell™ 16 FFPE Plus LEV DNA Purification Kit
The Maxwell 16 FFPE Plus LEV DNA Purification Kit is designed to extract DNA rapidly from FFPE samples using the Maxwell 16 Instrument (Cat.# AS2000 and AS3000).
Supplier: Promega AS1135
Description
- Fast, Automated DNA Extraction from FFPE Tissues
- Purifies DNA from 1-10 (5μm) FFPE samples
- Faster digestion option without organic solvents
- Sufficient yield for downstream amplification
Specifications
| 15°C to 30°C | |
| Extract DNA rapidly from FFPE samples | |
| 48 Preps |
| Maxwell™ 16 Instrument | |
| 48 Maxwell™ 16 LEV RNA cartridge, 50 LEV plungers, 50 0.5mL Elution tubes, 25mL Nuclease-free Water, 32mL Lysis Buffer, 15mL Incubation Buffer, 2 Proteinase K tubes |