missing translation for 'onlineSavingsMsg'
Learn More

Promega Mass Spec Grade Trypsin/Lys-C Mix

Catalog No. PRV5073
Encompass
Change view
Click to view available options
Quantity:
1 x 100 μg
1 x 20 μg
5 x 20μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV5073 5 x 20μg
PRV5071 1 x 20 μg
PRV5072 1 x 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PRV5073 Supplier Promega Supplier No. V5073

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Trypsin/Lys-C Mix, Mass Spec Grade, is a mixture of Trypsin Gold, Mass Spectrometry Grade, and rLys-C, Mass Spec Grade. The Trypsin/Lys-C Mix is designed to improve digestion of proteins or protein mixtures in solution.

  • Improve Protein Digestion with this Mix of Trypsin Gold and rLys-C
  • Use standard overnight digestion, nondenaturing conditions
  • Increased peptide recovery, enhanced protein quantitation, improved reproducibility
  • Tolerant to trypsin-inhibiting contaminants

Specifications

Color Colorless
Components 5 x 20μg Mass spec grade lyophilized Trypsin/Lys-C Mix, 500μL Resuspension Buffer
For Use With (Application) In-solution Protein digestion, in-solution digestion of proteolytically resistant Proteins
Storage Buffer 50mM Acetic Acid (pH <3) Resuspension Buffer
Content And Storage -30°C to -10°C
Quantity 5 x 20μg
Product Type Trypsin/Lys-C Mix
Form Liquid