Learn More
Promega Mass Spec Grade Trypsin/Lys-C Mix
Trypsin/Lys-C Mix, Mass Spec Grade, is a mixture of Trypsin Gold, Mass Spectrometry Grade, and rLys-C, Mass Spec Grade. The Trypsin/Lys-C Mix is designed to improve digestion of proteins or protein mixtures in solution.
Supplier: Promega V5072
Description
- Improve Protein Digestion with this Mix of Trypsin Gold and rLys-C
- Use standard overnight digestion, nondenaturing conditions
- Increased peptide recovery, enhanced protein quantitation, improved reproducibility
- Tolerant to trypsin-inhibiting contaminants
Chemical Identifiers
| Colorless | |
| 50mM Acetic Acid (pH <3) Resuspension Buffer | |
| Trypsin/Lys-C Mix |
| In-solution Protein digestion, in-solution digestion of proteolytically resistant Proteins | |
| -30°C to -10°C | |
| Liquid |
Specifications
| Colorless | |
| In-solution Protein digestion, in-solution digestion of proteolytically resistant Proteins | |
| -30°C to -10°C | |
| Trypsin/Lys-C Mix |
| 100μg Mass spec grade lyophilized Trypsin/Lys-C Mix, 500μL Resuspension Buffer | |
| 50mM Acetic Acid (pH <3) Resuspension Buffer | |
| 1 x 100 μg | |
| Liquid |