Learn More
Promega Mass Spec Grade Trypsin/Lys-C Mix
Trypsin/Lys-C Mix, Mass Spec Grade, is a mixture of Trypsin Gold, Mass Spectrometry Grade, and rLys-C, Mass Spec Grade. The Trypsin/Lys-C Mix is designed to improve digestion of proteins or protein mixtures in solution.
Supplier:  Promega V5071
Encompass
 View more versions of this product
Catalog No. PRV5071
Only null left
Description
Description
  • Improve Protein Digestion with this Mix of Trypsin Gold and rLys-C
  • Use standard overnight digestion, nondenaturing conditions
  • Increased peptide recovery, enhanced protein quantitation, improved reproducibility
  • Tolerant to trypsin-inhibiting contaminants
Specifications
Chemical Identifiers
Colorless
50mM Acetic Acid (pH <3) Resuspension Buffer
Trypsin/Lys-C Mix
In-solution Protein digestion, in-solution digestion of proteolytically resistant Proteins
-30°C to -10°C
Liquid
Specifications
Colorless
In-solution Protein digestion, in-solution digestion of proteolytically resistant Proteins
-30°C to -10°C
Trypsin/Lys-C Mix
20μg Mass spec grade lyophilized Trypsin/Lys-C Mix, 500μL Resuspension Buffer
50mM Acetic Acid (pH <3) Resuspension Buffer
1 x 20 μg
Liquid
Product Suggestions
Product Suggestions