Learn More
Promega Luciferase Control RNA DFS Item
Luciferase Control RNA is an uncapped in vitro-transcribed RNA containing a 30-base poly(A) tail that produces functional luciferase when translated.
Supplier:  Promega L4561
Catalog No. PR-L4561
Only null left
Description
Description
  • Unique Functional Control to Monitor In Vitro Translation Reactions
  • Control reactions are easily monitored using a luciferase assay
  • Safe, non-radioactive format to monitor control activity
  • Supplied at a concentration of 1mg/ml
  • The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713; and 5,700,673. A license (from Promega for research reagent products and from The Regents of the University of California for all other fields) is needed for any commercial sale of nucleic acid contained within or derived from this product.
  • Australian Patents 616881 and 641261 and other pending and issued patents, which claim vectors encoding a portion of human placental ribonuclease inhibitor, are exclusively licensed to Promega Corporation.
Specifications
Specifications
1 mg/mL
<-65°C
20 μg
7
For in-vitro translation reactions
Product Suggestions
Product Suggestions