missing translation for 'onlineSavingsMsg'
Learn More

Promega Luciferase Assay Systems

Catalog No. p-121409
Encompass
Change view
Click to view available options
Quantity:
100 Assays
1000 Assays
Product Type:
Luciferase Assay System
Luciferase Assay System Freezer Pack
Luciferase Assay System w/Reporter Lysis Buffer
Luciferase Assay System, 10-pack
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
PRE1500 100 Assays Luciferase Assay System
PRE4030 100 Assays Luciferase Assay System w/Reporter Lysis Buffer
PRE4530 1000 Assays Luciferase Assay System Freezer Pack
PRE4550 1000 Assays Luciferase Assay System
PRE1501 1000 Assays Luciferase Assay System, 10-pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. PRE1500 Supplier Promega Supplier No. E1500
Only null left

An extremely sensitive reagent for rapid quantitation of firefly luciferase. Can be used for assays in single-tube luminometers or in multiwell-plate luminometers with auto-injectors.

  • Rapid Detection of Firefly Luciferase Activity
  • Linear over eight or more orders of magnitude of enzyme concentration.
  • Sensitive to 10-20 moles of luciferase.
  • Cell lysis, sample prep and assay complete in as little as 5 minutes

  • U.S. Patents 5,641,641; 5,650,289; and 5,814,471; Australian Patent 649289 and other patents and patents pending.
  • The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713; and 5,700,673.
  • Certain applications of this product may require licenses from others.

Specifications

Product Type Luciferase Assay System
Includes 1 vial Luciferase Assay Substrate, 10mL Luciferase Assay Buffer, 30mL Cell Culture Lysis 5X Reagent
pH 4.6
Content And Storage -30°C to -10°C
Quantity 100 Assays
Form Liquid