missing translation for 'onlineSavingsMsg'
Learn More

Promega LCK Kinase Enzyme System

Catalog No. PRV2691
Encompass
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV2691 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRV2691 Supplier Promega Supplier No. V2691

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Recombinant full-length human LCK was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. LCK (p56lck) is a member of the src family of nonreceptor tyrosine kinases.

  • Easily Screen and Profile LCK Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Specifications

Content And Storage <-65°C
For Use With (Application) Indicates initiation and maintenance of transformed state in Human cancer
Quantity 10 μg