missing translation for 'onlineSavingsMsg'
Learn More

Promega ITK Kinase Enzyme System

Catalog No. PRV9431
Encompass
Change view
Click to view available options
Quantity:
1/Ea.
10 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV9431 1/Ea.
PRV3191 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV9431 Supplier Promega Supplier No. V9431

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

ITK Kinase Enzyme System (V3191)

  • Kinase: ITK, 10μg (Human, recombinant; amino acids 352–end), M.W.: ∽53kDa
  • Substrate: Poly (4:1 Glu, Tyr) Peptide
  • Others: Reaction Buffer, DTT, MnCl2
ADP-Glo™ Kinase Assay and ITK Kinase Enzyme System (V9261)
  • 1 x 10μg ITK Kinase Enzyme System (V3191) and 1 x 1000 ADP-Glo™ Kinase Assays (V9261)

Recombinant human ITK (amino acids 352-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. ITK is a member of the TEC family of nonreceptor tyrosine kinases.

  • Easily Screen and Profile ITK Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Specifications

Molecular Weight (g/mol) 53
Content And Storage Components at different temps
Quantity 1/Ea.
Form Fluid