missing translation for 'onlineSavingsMsg'
Learn More

Promega IGF1R Kinase Enzyme System

Catalog No. PRV9401
Encompass
Change view
Click to view available options
Quantity:
1/Ea.
10 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV9401 1/Ea.
PRV3581 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV9401 Supplier Promega Supplier No. V9401

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

IGF1R Kinase Enzyme System (V3581)

  • Kinase: IGF1R, 10μg (Human, recombinant; amino acids 960–end). MW: ∽53kDa
  • Substrate: IGF1Rtide (KKKSPGEYVNIEFG); derived from human IRS-1 protein residues 891–902
  • Reaction Buffer, DTT, MnCl2
ADP-Glo™ Kinase Assay and EGFR Kinase Enzyme System (V9401)
  • 1 x 10μg EGFR Kinase Enzyme System (V3581) and 1 x 1000 ADP-Glo™ Kinase Assays (V9101)

Recombinant human IGF1R was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag. IGF1R (Insulin-like Growth Factor 1) is a transmembrane tyrosine kinase receptor that is activated by IGF-1 and by the related growth factor IGF-2.

  • Easily Screen and Profile IGF1R Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Specifications

Includes 1 x 10μg EGFR Kinase Enzyme system and 1 x 1000 ADP-Glo™ Kinase assay
Molecular Weight (g/mol) 53
Content And Storage Components at different temps
For Use With (Application) Growth and anabolic effects in adults
Quantity 1/Ea.
Form Fluid