missing translation for 'onlineSavingsMsg'
Learn More

Promega E. coli S30 T7 High-Yield Protein Expression System

Catalog No. PRL1115
Encompass
Change view
Click to view available options
Quantity:
24 reactions
8 reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRL1115 8 reactions
PRL1110 24 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRL1115 Supplier Promega Supplier No. L1115

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

T7 S30 Extract, an optimized S30 Premix Plus provides all other required components, including amino acids, rNTPs, tRNAs, an ATP-regenerating system, IPTG and appropriate salts to express high levels of recombinant proteins. Also includes S30 T7 Control DNA, a control DNA template.

The E. coli S30 T7 High-Yield Protein Expression System is designed to express up to 500ug/ml of protein in 1 hour from plasmid vectors containing a T7 promoter and a ribosome binding site.

  • Simplified Transcription/Translation of DNA Cloned in Plasmid or Lambda Vectors with a T7 Promoter
  • Express up to 500μg/mL of protein in one hour
  • Flexible detection of expressed proteins by Coomassie(R) staining or fluorescence or biotinylated labels

Specifications

Test Time 1 hr.
Yield 500 μg/mL
Type E. coli Protein Expression System
Sufficient For 8 Reactions
Quantity 8 reactions
Content And Storage <-65°C
For Use With (Application) Protein-Protein interactions, Protein-DNA interactions, Enzyme activity, Antibody production

U.S. Patents 5,292,658, and 5,418,155; Canadian Patent 2,105,984 and related patents.