missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega HisLink™ Protein Purification Resins
HisLink Protein Purification Resin is a macroporous silica resin modified to contain a high level of tetradentate-chelated nickel (>20mmol Ni/ml settled resin) for efficient capture and purification of bacterially expressed His- or HQ-tagged proteins.
$97.51 - $642.99
Specifications
| Content And Storage | 2°C to 10°C |
|---|---|
| For Use With (Application) | Purification of His-tagged Proteins from cleared or crude bacterial lysates |
| Type | Resin |
Description
- Resin to Purify His- or HQ-Tagged Protein from Cleared Lysate
- Purify polyhistidine- or HQ-tagged proteins from cleared lysate by standard column chromatography or from crude lysate in batch
- Suitable for low- to medium-pressure applications such as FPLC
- Large quantities of polyhistidine- or HQ-tagged protein can be purified per volume of resin used
- U.S. Patent No. 7,112,552 and other patents pending.
- Certain applications of this product are covered by patents issued and applicable in certain countries. Because purchase of this product does not include a license to perform any patented application, users of this product may be required to obtain a patent license depending upon the particular application and country in which the product is used.
Specifications
| 2°C to 10°C | |
| Purification of His-tagged Proteins from cleared or crude bacterial lysates | |
| Resin |