Learn More
Promega HER4 Kinase Enzyme System
Description
Includes
HER4 Kinase Enzyme System (V3101)
- Kinase: HER4, 10μg (Human, recombinant; amino acids 682–993). MW: ∽57kDa
- Substrate: Poly (4:1 Glu, Tyr) Peptide.
- Others: Reaction Buffer, DTT, MnCl2
- 1 x 10μg EGFR Kinase Enzyme System (V3101) and 1 x 1000 ADP-Glo™ Kinase Assays (V9101)
- Easily Screen and Profile HER4 Kinase Inhibitors
- Includes kinase, substrate and reaction buffer
- Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Specifications
| Includes | 1 x 10μg HER4 Kinase Enzyme system, 1 x 1000 assays ADP-Glo™ Kinase assay |
| Content And Storage | Components at different temps |
| Quantity | 1/Ea. |