missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega HER2 Kinase Enzyme System
Description
- Easily Screen and Profile HER2 Kinase Inhibitors
- Includes kinase, substrate and reaction buffer
- Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Specifications
| Molecular Weight (g/mol) | 116 |
| Content And Storage | -70°C |
| For Use With (Application) | Cellular transformation and tumorigenesis |
| Quantity | 1000 Assays |
| Form | Fluid |