Learn More
Promega HB101 Competent Cells
HB101 cells are recA- to minimize recombination events and contain a restriction-minus background (hsdS20) to allow better representation when cloning methylated DNA. Suitable for vectors that do not require alpha-complementation for blue/white screening.
Supplier: Promega L2015
Description
- Ready-to-Use Competent E. coli Cells for Recombinant Cloning and Subcloning
- Cells are ready to use—no preparation time necessary
- Transformation efficiencies guaranteed
- No worries about undesirable recombination events (recA- mutation)
Specifications
| >108cfu/μg | |
| Transformation of unmethylated DNA derived from PCR, cDNA and many other sources | |
| 7 | |
| 20 x 50 μL |
| Store at -70°C. | |
| Single-Use HB101 Competent Cells | |
| Competent Cells | |
| Fluid |