missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega GSK3α Kinase Enzyme System
Description
Includes
- 1 x 10μg GSK3α Kinase Enzyme System (V3051) and 1 x 1000 ADP-Glo™ Kinase Assays (V9101)
- Easily Screen and Profile GSK3α Kinase Inhibitors
- Includes kinase, substrate and reaction buffer
- Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Specifications
| Includes | 1 x 10μg GSK3a Kinase Enzyme system, 1 x 1000 ADP-Glo™ Kinase assay |
| Molecular Weight (g/mol) | 81 |
| Content And Storage | Components at different temps |
| Quantity | 1/Ea. |
| Form | Fluid |