Learn More
Promega GSK3β Kinase Enzyme System
Description
Includes
GSK3β Kinase Enzyme System (V1991)
- Kinase: GSK3β, 10μg (Human, recombinant full length). MW: ∽73kDa
- Substrate: GSK3 derived from human muscle glycogen synthase 1 (amino acid 636–661)
- Others: Reaction Buffer, DTT, MnCl
- 1 x 10μg GSK3β Kinase Enzyme System (V1991) and 1 x 1000 ADP-Glo™ Kinase Assays (V9101)
- Easily Screen and Profile GSK3β Kinase Inhibitors
- Includes kinase, substrate and reaction buffer
- Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Specifications
| Includes | 1 x 10μg GSK3beta Kinase Enzyme system, 1 x 1000 assays ADP-Glo™ Kinase assay |
| Content And Storage | Components at different temps |
| Quantity | 1/Ea. |