missing translation for 'onlineSavingsMsg'
Learn More

Promega GoTaq™ Hot Start Polymerase: Green Master Mix, 2X

Catalog No. PRM5123
Encompass
Change view
Click to view available options
No. of Reactions:
100
1000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
PRM5123 1000
PRM5122 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRM5123 Supplier Promega Supplier No. M5123
Only null left

Includes

GoTaq Hot Start Polymerase, buffer, dNTPs, and MgCl2, and supplied with nuclease-free water.

GoTaq™ Hot Start Master Mixes are all-in-one master mixes (just add template and primers) that contain the high-performance GoTaq™ DNA Polymerase bound to a proprietary antibody that blocks polymerase activity until the reaction is heated at 94-95°C for two

  • Hot-Start GoTaq(R) DNA Polymerase in a Ready-to-Use Master Mix
  • All-in-one master mix: just add template and primers
  • Minimize nonspecific amplification and primer-dimers
  • Use the green master mix for direct-to-gel analysis, and the colorless master mix when post-amplification fluorescence or absorbance analysis is required

  • U.S. Patent No. 6,242,235, Australian Patent No. 761757, Canadian Patent No. 2,335,153.
  • Licensed under U.S. Patent Nos. 5,338,671 and 5,587,287 and corresponding patents in other countries.

Specifications

Concentration 5 U/μL
Content And Storage 2X Hot start green master mix (1 x 25mL), Nuclease free Water (1 x 25mL). Store at -20°C
For Use With (Application) Cloning
Activation Time 2 min.
No. of Reactions 1000