missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega GoTaq™ Hot Start Polymerase: 5X
Description
Includes
GoTaq Hot Start Polymerase is supplied with 5X Colorless GoTaq Flexi Buffer, 5X Green GoTaq Flexi Buffer, and 25mM MgCl2.
- Robust PCR with Convenient Room-Temperature Setup
- Minimize nonspecific amplification and primer-dimers
- Optimize reaction conditions with supplied Flexi Reaction Buffer and MgCl2 solution
- Direct-to-gel convenience
- Supplied at a concentration of 5u/μL
- U.S. Patent No. 6,242,235, Australian Patent No. 761757, Canadian Patent No. 2,335,153.
- Licensed under U.S. Patent Nos. 5,338,671 and 5,587,287 and corresponding patents in other countries.
Specifications
Specifications
| Concentration | 5 U/μL |
| Content And Storage | 5X Hot start Polymerase (1 x 100U), colorless flexi Buffer (1 x 1mL), 5X green flexi Buffer (1 x 1mL), MgCl2 (1 x 750μL). Store at −30 to −10°C |
| Quantity | 100U |
| For Use With (Application) | Cloning |
| Activation Time | 2 min. |