missing translation for 'onlineSavingsMsg'
Learn More

Promega GoTaq™ 2-Step RT-qPCR System

Catalog No. PRA6010
Encompass
Change view
Click to view available options
Quantity:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRA6010 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRA6010 Supplier Promega Supplier No. A6010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

GoTaq QPCR Master Mix, 2X; GoScript Reverse Transcriptase; GoScript Reaction Buffer, 5X; CXR Reference Dye, 100X; Recombinant RNasin Ribonuclease Inhibitor; Random Primers; PCR Nuceotide Mix; Oligo (dT)15; MgCl2; Nuclease-free Water

Combines the ultra-active GoScript™ Reverse Transcriptase with the ultra-bright fluorescence of GoTaq qPCR Master Mix to provide optimized, sensitive quantification of a full range of RNA targets

  • Convenient Master Mixes for Quantitative Real-Time PCR and RT-qPCR
  • Ready-to-use mix of optimized components for dye-based qPCR or RT-qPCR
  • Compatible with both fast and standard cycling methods
  • Resistant to a wide range of PCR inhibitors

Specifications

Content And Storage 2X QPCR Master mix (5 x 1mL), GoScript™ reverse Transcriptase (1 x 50μL), 5X GoScript™ reaction Buffer (1 x 300μL), 100X CXR Reference Dye (1 x 100μL), Recombinant R. Store at −30 to −10°C
Detection Method qPCR, RT-PCR
Form Liquid
Format Kit
PCR Method 2-step RT-qPCR
Polymerase GoTaq™ Hot Start Polymerase
Technique RT-qPCR, Reverse Transcription
DNase DNase-free
For Use With (Equipment) RT- PCR Instrumen Detecting SYBR Green I or FAM Dye
High-throughput Compatibility High-throughput Compatible
Includes Label or Dye Yes
Kit Type 2-Step RT-qPCR system
Label or Dye Fluorescein (FITC)
No. of Tests 50
Passive Reference Dye CXR (Separate Tube)
Product Line GoTaq, GoScript
Product Type RT-qPCR Kit
Quantity 5mL
Research Category Gene Expression
Reverse Transcriptase GoScript™
Ribonuclease H Activity Yes
Shipping Condition Dry Ice
Sufficient For 50 RT Reactions and 500 x 20 μL qPCR Reactions
For Use With (Application) Gene Expression
Activation Time 2 min.
Compatibility RT-PCR Instruments that Detect SYBR Green I or FAMTM Dye
Final Product Type First-Strand cDNA
Hot Start Built-In Hot Start
No. of Reactions 500 Reactions
Reagent Type RT-qPCR, cDNA Synthesis
Sample Type Blood, Cells, Feces, FFPE, Food, Human Cerebrospinal Fluid, Plants, Plasma, Saliva, Serum, Sputum, Swabs, Tissue, Urine
Starting Material RNA
Volume 20 μL
Storage Buffer 2X GoTaq™ qPCR Master Mix, 5X GoScript™ Reaction Buffer
Show More Show Less

  • U.S. Patent 6,242,235, Australian Patent 761757, Canadian Patent 2,335,153 and other patents and patents pending.
  • U.S. Patent 5,552,302.
  • Licensed under U.S. Patents 5,338,671 and 5,587,287 and corresponding patents in other countries. For research use only. Not for use in diagnostic procedures.
  • Notice to Purchaser: Limited License
  • Use of this product is covered by one or more of the following U.S. patents and corresponding patent claims outside the U.S.: 5,789,224; 5,618,711; 6,127,155 and claims outside the U.S. corresponding to U.S. Patent 4,889,818. The purchase of this product includes a limited, nontransferable immunity from suit under the foregoing patent claims for using only this amount of product for the purchaser's own internal research. No right under any other patent claim (such as the patented 5' Nuclease Process claims in U.S. Patents 5,210,015 and 5,487,972), no right to perform any patented method, and no right to perform commercial services of any kind, including without limitation reporting the results of purchaser's activities for a fee or other commercial consideration, is conveyed expressly, by implication, or by estoppel. This product is for research use only. Diagnostic uses under Roche patents require a separate license from Roche. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404 USA.