missing translation for 'onlineSavingsMsg'
Learn More

Promega GoTaq™ 1-Step RT-qPCR System for Dye-Based Detection

Catalog No. PRA6020
Encompass
Change view
Click to view available options
Quantity:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRA6020 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRA6020 Supplier Promega Supplier No. A6020

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For quantitative analysis of RNA using a one-step reverse transcription-quantitative PCR (RT-qPCR) protocol for dye-based detection.

  • Convenient Master Mixes for Quantitative Real-Time PCR and RT-qPCR
  • Ready-to-use mix of optimized components for dye-based qPCR or RT-qPCR
  • Compatible with both fast and standard cycling methods
  • Resistant to a wide range of PCR inhibitors

Specifications

Content And Storage 2X qPCR Master mix (5 x 1mL), RT Mix (1 x 225μL), CXR reference Dye (1 x 200μL), MgCl2 (1 x 750μL), Nuclease free Water (2 x 13mL). Store at −30 to −10°C
Detection Method SYBR
Form Liquid
Format Kit
PCR Method 1-step RT-qPCR
Polymerase GoTaq™ DNA Polymerase
Reaction Speed Fast or Standard
Technique RT-qPCR
For Use With (Equipment) Analytik Jena qTOWERiris PCR thermal cycler
High-throughput Compatibility High-throughput Compatible
Includes Label or Dye Yes
Kit Type 1-Step RT-qPCR system
Label or Dye SYBR Green
No. of Tests 200
Passive Reference Dye CXR (Separate Tube)
Product Line GoTaq
Product Type RT-qPCR Kit
Quantity 5mL
Research Category Gene Expression Analysis
Reverse Transcriptase GoScript™
Sufficient For 500 Reactions of 20 μL
For Use With (Application) Real Time PCR (qPCR)
Final Product Type cDNA
Hot Start Built-In Hot Start
No. of Reactions 500 Reactions
Reaction Format Separate Components
Reagent Type RT-qPCR, Reverse Transcription
Sample Type Blood
Starting Material RNA
Show More Show Less