Learn More
Promega Go Taq™ Master Mixes
New formulation of Taq DNA Polymerase with enhanced performance over standard Taq DNA Polymerase. Available in two all-in-one master mixes: green and colorless. The green master mix allows direct gel analysis after amplification.
Supplier: Promega M7132
Includes: GoTaq Green Master Mix or GoTaq Colorless Master Mix, Nuclease-free water
Description
- All-in-One Optimized PCR Master Mix-Just Add Template, Primers and Water
- Directly substitute into your current PCR conditions
- Use the green master mix for direct-to-gel analysis after amplification
- Use the colorless master mix for post-amplification analysis by fluorescence or absorbance without prior DNA purification
Chemical Identifiers
| 2X | |
| Standard PCR |
Specifications
| 2X | |
| Go Taq™ Colorless Master Mix | |
| 100 Reactions |
| • 2X GoTaq™ Colorless Master Mix, 2 x 1.25 mL • Nuclease-free Water, 2 x 1.25 mL Store at –30° to ‐10°C. |
|
| Standard PCR |