missing translation for 'onlineSavingsMsg'
Learn More

Promega Glycosidases

Catalog No. PRV4875
Encompass
Change view
Click to view available options
Quantity:
10,000 U
50,000 U
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV4875 50,000 U
PRV4871 10,000 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV4875 Supplier Promega Supplier No. V4875

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Endo H is cloned from S. plicatus and overexpressed in E. coli.

  • Characterize Protein Glycosylation
  • Monitor protein trafficking
  • Characterize glycan location and structure
  • Does not cleave complex glycans
  • Supplied at a concentration of 500u/μL

Specifications

Concentration 500 U/μL
For Use With (Application) Determining the state of Protein Glycosylation, characterizing Glycan structure
Formulation 1 x 50000U Endo H, 1 x 1mL 10X Endo H Reaction Buffer, 1 x 1mL 10X Denaturing Solution
Content And Storage -30°C to -10°C
Quantity 50,000 U
Product Type Endoglycosidase H