missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega Glu-C Sequencing Grade Serine Protease
Description
- Cleaves at the C-Terminus of Either Aspartic or Glutamic Acid Residues
- Optimal activity at pH 4-9
- Ammonium and phosphate buffers can affect cleavage; see details in product information
- Suitable for in-solution digestion; not recommended for in-gel digestion
Specifications
Specifications
| Color | Colorless |
| pH | 4 to 9 |
| For Use With (Application) | Cleaves at the C-terminus of either Aspartic or Glutamic Acid residues (1 to 4) |
| Content And Storage | -30°C to +10°C |
| Source | S. aureus V8 |
| Quantity | 50 μg |
| Product Type | Glu-C Serine Protease |
| Molecular Weight (g/mol) | 27 |
| Inhibitors | Diisopropyl Fluorophosphate (DFP) Monovalent anions such as F-, Cl- Br-, CH3COO- and NO3- (1,5) |
| Form | Lyophilized Liquid |