Learn More
Promega pF3 WG (BYDV) Flexi™ Vectors
Designed for use with Wheat Germ extracts and contain sequences from the barley yellow dwarf virus (BYDV), an RNA plant virus, upstream and downstream of the protein coding region of interest.
Supplier: Promega L5671
Description
- Protein Expression Vectors Used with Wheat Germ Cell-Free Systems
- Cloned inserts are in vitro translated in WGE systems
- Adaptable to high-throughput formats for large screening projects
- Transfer your cloned insert into other Flexi(R) Vectors for additional applications (e.g., bacterial or mammalian protein expression) as needed
Specifications
| 100ng/μL | |
| Inducible expression of HQ- and GST-tagged fusion Proteins in E. coli and cell-free systems using the T7 RNA polymerase promoter | |
| 8 | |
| 0.02 mg |
| -30°C to -10°C | |
| 10X Flexi Enzyme blend (Sgf I and Pme I), 2X flexi Ligase Buffer, 5X flexi Digest Buffer, Nuclease-free Water, T4 DNA Ligase, Wizard™ SV Gel and PCR clean up system (C8640 only) | |
| pF3A WG (BYDV) Flexi Vector | |
| Fluid |