Learn More
Promega Flexi™ Enzyme Blends
The 10X Flexi Enzyme Blend (SgfI and PmeI) is required to screen Flexi Vector clones for the presence of the protein-coding region of interest. This enzyme blend can also be used to cleave PCR products prior to cloning into the Flexi Vectors or transfer p
Supplier: Promega R1852
Description
- Reagent Used with Flexi(R) Cloning System
- Required to screen Flexi(R) Vector clones for presence of protein-coding region of interest
- Also can be used to cleave PCR products prior to cloning into Flexi(R) Vectors or transfer protein-coding regions between Flexi(R) Vectors
Specifications
| -30°C to -10°C | |
| 10X Flexi Enzyme blend (SgfI and Pmel)1 x 100μL, Nuclease-free Water 2 x 1,250μL, 5X Flexi Digest Buffer 1 x 1mL | |
| Enzyme Blend | |
| Fluid |
| Flexi cloning system | |
| 7.5 | |
| 100 μL |
Licensed under U.S. Patent 5,945,288.