Learn More
Promega FGFR1 Kinase Enzyme System
Description
- Easily Screen and Profile FGFR1 Kinase Inhibitors
- Includes kinase, substrate and reaction buffer
- Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Specifications
| Includes | 1 x 10μg FGFR1 Kinase Enzyme system, 1 x 1000 assays ADP-Glo™ Kinase assay |
| Content And Storage | Components at different temps |
| Quantity | 1/Ea. |