missing translation for 'onlineSavingsMsg'
Learn More

Promega FastBreak™ Cell Lysis Reagent

Catalog No. prv8571
Encompass
Change view
Click to view available options
Quantity:
15 mL
60 mL
100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV8571 15 mL
PRV8573 100 mL
PRV8572 60 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PRV8571 Supplier Promega Supplier No. V8571
Only null left

FastBreak Cell Lysis Reagent allows efficient, gentle lysis of E. coli cultures without the need for centrifugation or mechanical disruption.

  • Quick E. coli Cell Culture Lysis, No Centrifugation or Mechanical Disruption Required
  • Eliminate centrifugation and mechanical disruption
  • Simple, add-and-incubate protocol
  • Compatible with manual or robotic platforms

  • Patent Pending.
  • This product is licensed for use under U.S. Patent 6,174,704.
  • Certain applications of this product are covered by patents issued and applicable in certain countries. Because purchase of this product does not include a license to perform any patented application, users of this product may be required to obtain a patent license depending upon the particular application and country in which the product is used.

Specifications

Concentration 10X
Product Type Cell Lysis Reagent
Content And Storage 4°C to 25°C
Quantity 15 mL