missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega FastBreak™ Cell Lysis Reagent
Description
- Quick E. coli Cell Culture Lysis, No Centrifugation or Mechanical Disruption Required
- Eliminate centrifugation and mechanical disruption
- Simple, add-and-incubate protocol
- Compatible with manual or robotic platforms
- Patent Pending.
- This product is licensed for use under U.S. Patent 6,174,704.
- Certain applications of this product are covered by patents issued and applicable in certain countries. Because purchase of this product does not include a license to perform any patented application, users of this product may be required to obtain a patent license depending upon the particular application and country in which the product is used.
Specifications
Specifications
| Concentration | 10X |
| Product Type | Cell Lysis Reagent |
| Content And Storage | 4°C to 25°C |
| Quantity | 100 mL |