Learn More
Promega EPHA1 Kinase Enzyme System
Description
- Easily Screen and Profile EPHA1 Kinase Inhibitors
- Includes kinase, substrate and reaction buffer
- Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Specifications
| Includes | 1 x 10μg EPHA1 Kinase Enzyme system, 1 x 1000 assays ADP-Glo™ Kinase assay |
| Content And Storage | Store the Kinase at -70°C and the rest of the components at -20°C |
| Quantity | Each |