missing translation for 'onlineSavingsMsg'
Learn More

Promega Eluator™ Vacuum Elution Device

Catalog No. PRA1071
Encompass
Change view
Click to view available options
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRA1071 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRA1071 Supplier Promega Supplier No. A1071
Only null left

Includes

Blue base and clear column assembly

The Eluator Vacuum Elution Device is used with PureYield Purification Systems to elute purified DNA or RNA without the need for centrifugation.

  • Use with PureYield™ Purification Systems to Elute Without Centrifugation
  • Increased DNA or RNA recovery compared to centrifugation protocols
  • Faster purification

Specifications

Content And Storage 15°C to 30°C
For Use With (Equipment) PureYield™ Midiprep/Maxiprep columns
For Use With (Application) Elute Nucleic Acids
Includes Blue base and clear column assembly
Product Type Accessory
Quantity Each