missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega EGFR Kinase Enzyme System
Description
Includes
EGFR Kinase Enzyme System (V3831)
- Kinase: EGFR, 10μg (Human, recombinant; amino acids 695–end). MW: ∽89kDa
- Substrate: Poly (4:1 Glu, Tyr) Peptide
- Others: Reaction Buffer, DTT, MnCl2
- 1 x 10μg EGFR Kinase Enzyme System (V3831) and 1 x 1000 ADP-Glo™ Kinase Assays (V9101)
- Easily Screen and Profile EGFR Kinase Inhibitors
- Includes kinase, substrate and reaction buffer
- Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Specifications
| Molecular Weight (g/mol) | 89 |
| Content And Storage | <-65°C |
| Quantity | 10 μg |
| Form | Fluid |