missing translation for 'onlineSavingsMsg'
Learn More

Promega Dual-Glo™ Luciferase Assay System

Catalog No. p-319429
Encompass
Change view
Click to view available options
Quantity:
10 mL
100 mL
10 x 100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRE2920 10 mL
PRE2940 100 mL
PRE2980 10 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PRE2920 Supplier Promega Supplier No. E2920
Only null left

Includes

Dual-Glo Luciferase Buffer; Dual-Glo Luciferase Substrate; Dual-Glo Stop & Glo™ Buffer; Dual-Glo Stop & Glo Substrate

A homogeneous reagent system that enables fast, simple quantitation of both firefly and Renilla luciferase luminescence signals from cells that have not been preconditioned or prelysed.

  • Detects firefly and Renilla Luciferase Activities in a Single Sample
  • Renilla luciferase internal control provides more accurate results
  • Assay cells directly in growth medium for both reporters
  • Extended half-life of 2 hours for each reporter

  • U.S. Patents 5,641,641; 5,650,289; and 5,814,471; Australian Patent 649289 and other patents.
  • U.S. Patent 5,744,320; Australian Patent 721172; Canadian Patent 2,221,522 and other patents.
  • U.S. Patents 7,078,181; 7,108,996; 7,118,878; Australian Patent 2001275325 and other patents pending.
  • The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713; and 5,700,673.
  • Certain applications of this product may require licenses from others.
  • This product does not convey a license to use recombinant Renilla luciferase under U.S. Patent 5,418,155 and related patents. Promega sells licensed Renilla luciferase vectors, which may be used in conjunction with this product.

Specifications

Content And Storage -30°C to -10°C
Format 96 and 384-well Plates
Quantity 10 mL