missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega CSK Kinase Enzyme System
Description
Includes
- 1 x 10μg CSK Kinase Enzyme System (V2981) and 1 x 1000 ADP-Glo™ Kinase Assays (V9101)
- Easily Screen and Profile CSK Kinase Inhibitors
- Includes kinase, substrate and reaction buffer
- Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Specifications
| Includes | 1 x 10μg CSK Kinase Enzyme system, 1 x 1000 assays ADP-Glo™ Kinase assay |
| Content And Storage | Components at different temps |
| For Use With (Application) | Ideal for both primary screening as well as Kinase selectivity profiling |
| Quantity | 1/Ea. |