missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega CHK1 Kinase Enzyme System
Description
Includes
- Kinase: CHK1, 10μg (Human, recombinant full length). MW: ∽82kDa
- Substrate: CHKtide (KKKVSRSGLYRSPSMPENLNRPR); derived from the human CDC25C protein isoform A (amino acid 205-225)
- Others: Reaction Buffer, DTT
- Easily Screen and Profile CHK1 Kinase Inhibitors
- Includes kinase, substrate and reaction buffer
- Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Specifications
| Includes | 1 x 10μg CHK1 Kinase Enzyme system, 1 x 1000 ADP-Glo™ Kinase assay |
| Molecular Weight (g/mol) | 82 |
| Content And Storage | Components at different temps |
| Quantity | 1/Ea. |
| Form | Fluid |