missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega cAMP-Glo™ Max Assay System
Description
Includes
- Protein Kinase A (V135A) : 1 x 20μL
- cAMP-Glo™ ONE Buffer (V166A) : 1 x 2.5mL
- MgCl2, 1M (V330A) : 1 x 1mL
- cAMP, 1mM (V642B) : 1 x 100μL
- Kinase-Glo™ Substrate (V672A) : 1 x 1 vial
- Kinase-Glo™ Buffer (V673A) : 1 x 10mL
- Faster cAMP Detection for High-Throughput Assays
- Complete in ∼30 minutes
- Scalable to 1536-well plate formats and beyond
- No interference by fluorescent compounds
Specifications
Specifications
| Includes | 1 x 20μL Protein Kinase A, 1 x 2.5mL cAMP-Glo™ one Buffer, 1 x 1mL 1M MgCl2, 1mM 1 x 100μL cAMP, Kinase-Glo™ substrate 1 x 1 Vial, 1 x 10mL Kinase-Glo™ Buffer |
| pH | 7.5 |
| Content And Storage | -30°C to -10°C |
| For Use With (Application) | High-throughput analysis of library compounds to identify modulators of GPCR activity, easy quantitation of cAMP levels in broad range of cell types |
| Quantity | 10 x 20 Plates |