missing translation for 'onlineSavingsMsg'
Learn More

Promega cAMP-Dependent Protein Kinase, Catalytic Subunit

Catalog No. PRV5161
Encompass
Change view
Click to view available options
Quantity:
2500 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV5161 2500 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRV5161 Supplier Promega Supplier No. V5161

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

May be used to phosphorylate target proteins or for in vitro enzymological studies of neural and hormonal signal transduction

  • Purified Kinase for Protein Phosphorylation and Signal Transduction Studies
  • Purified from recombinant E. coli expressing the catalytic subunit of bovine PKA; 90% pure
  • Supplied at a concentration of 1.5-3mg/ml

Specifications

Concentration 1.5 to 3 mg/mL
Color Colorless
Purity 90%
pH 6.5
For Use With (Application) Phosphorylate target Proteins or for in-vitro enzymological studies of neural and hormonal signal transduction
Storage Buffer 350mM Potassium Phosphate (pH 6.8), 0.1mM DTT
Content And Storage <-65°C
Source Recombinant E. coli strain
Quantity 2500 U
Product Type cAMP-dependent Protein Kinase
Molecular Weight (g/mol) 40
Inhibitors AMP-dependent Protein Kinase Peptide
Form Liquid
Show More Show Less