Learn More
Promega BL21(DE3)pLysS Competent Cells
BL21(DE3)pLysS Competent Cells allow high-efficiency protein expression of any gene that is under the control of a T7 promoter and has a ribosome binding site.
Supplier: Promega L1195
Description
- High-Efficiency Protein Expression from Gene with T7 Promoter and Ribosome Binding Site
- Inducible protein expression from gene with T7 RNA polymerase controlled by lac UV5 promoter
- Increased stability of expressed protein from deficiency in proteases lon and OmpT
- Lower background expression of target genes with pLysS plasmid
Specifications
| <-65°C | |
| Allow high-efficiency Protein expression of genes under the control of a T7 promoter and having a ribosome binding site | |
| 20 x 50 μL |
| Tube | |
| Competent cell control DNA, BL21(DE3)pLysS competent cells >10 x 6cfu/μg |