missing translation for 'onlineSavingsMsg'
Learn More

Promega Asp-N Sequencing Grade Endoproteinase

Catalog No. PRV1621
Encompass
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV1621 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRV1621 Supplier Promega Supplier No. V1621

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Asp-N, Sequencing Grade, is an endoproteinase that hydrolyzes peptide bonds on the N-terminal side of aspartic and cysteic acid residues: Asp and Cys.

  • Hydrolyzes Peptide Bonds on the N-Terminal Side of Aspartic and Cysteic Acid Residues (Asp and Cys)
  • Optimal activity in pH range of 4-9
  • Sequencing grade enzyme for use alone or in combination with other proteases
  • Use for in-solution or in-gel digestion reactions

Specifications

Color Colorless
Purity ≥98%
pH 4 to 9
For Use With (Application) Hydrolyzing the Peptide on the N-terminal side of Aspartic and Cysteic Acid residues: Asp and Cys
Storage Buffer 10mM Tris HCl (pH 8)
Content And Storage 2°C to 10°C
Source Pseudomonas fragi
Quantity 2 μg
Product Type Endoproteinase
Molecular Weight (g/mol) 24.5
Form Lyophilized Powder
Show More Show Less