missing translation for 'onlineSavingsMsg'
Learn More

Promega Arg-C Sequencing Grade Modifying Enzyme

Catalog No. prv1881
Encompass
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV1881 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRV1881 Supplier Promega Supplier No. V1881
Only null left

Arg-C (clostripain) is an endopeptidase that cleaves at the C-terminus of arginine residues, including the sites next to proline. Cleavage also will occur at lysine residues. Arg-C activity is optimal in the pH range of 7.6-7.9.

  • Sequencing-Grade Protease: for use Alone or with other Proteases for Mass Spectrometry Analysis
  • Cleaves at the C-terminus of arginine residues, including sites next to proline
  • Also cleaves at lysine residues
  • Activity is optimal in the pH range of 7.6-7.9

Specifications

Color Colorless
pH 7.6 to 7.9
For Use With (Application) Peptide mapping, Protein identification, characterization of post-translational modifications
Storage Buffer 50mM Tris HCl (pH 7.6 to 7.9), 5mM CaCl2, 2mM EDTA
Content And Storage 2°C to 10°C
Source Clostridium histolyticum
Quantity 10 μg
Product Type Arg-C (clostripain)
Molecular Weight (g/mol) 45 and 12
Inhibitors Oxidizing agents and Sulfhydryl reactants, also Co2+, Cu2+, Cd2+ and heavy Metal Ions, Citrate, Borate and Tris anions partially inhibit activity, TLCK (Tosyllysine Chloromethyl Ketone) reacts with active site (1,5,6)
Form Lyophilized Powder
Show More Show Less