missing translation for 'onlineSavingsMsg'
Learn More

Promega 4-CORE™ Buffer Pack

Catalog No. PRR9921
Encompass
Change view
Click to view available options
Quantity:
4 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRR9921 4 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRR9921 Supplier Promega Supplier No. R9921
Only null left

Includes

Promega 10X restriction enzyme buffers A, B, C, D (1mL each)

Consists of one vial each of Restriction Enzyme Buffers A, B, C and D.

  • Aliquots of 10X Restriction Enzyme Buffers A, B, C and D
  • Majority of Promega restriction enzymes have optimal activity in one of these four 10X reaction buffers

Specifications

Color Colorless
Components 10X Restriction Enzyme Buffers A, B (pH 7.8), C (pH 8.2), D (pH 8.2) (1mL each) , 8.2
For Use With (Application) Pharmaceuticals
Content And Storage -30°C to -10°C
Quantity 4 x 1 mL
Product Type Buffer Pack
Form Liquid